Learn More
Medchemexpress LLC BOP sodium | 1947348-42-6 | 99.3% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1294531MG
Encompass_Preferred
BOP sodium is a potent and selective dual α9β1/α4β1 integrin inhibitor with Kd values in the picomolar range. It shows rapid and preferential mobilization of hematopoietic stem cell (HSC) and progenitors. It has little inhibitory activity on α4β7, α1β1, α2β1, and α5β1, αIIBβ3 integrins.
  • Potent and selective dual α9β1/α4β1 integrin inhibitor
  • Kd values in the picomolar range
  • Rapid and preferential mobilization of hematopoietic stem cells (HSC) and progenitors
  • Exhibits little inhibitory activity on α4β7, α1β1, α2β1, and α5β1, αIIBβ3 integrins
Catalog No. 50-003-40386