Learn More
Medchemexpress LLC Borate(1-), [5-[(3,5-dimethyl-2H-pyrrol-2-ylidene-KN)methyl]-1H-pyrrole-2-dodecanoato(2-)-KN1]difluoro-, hydrogen (1:1), (T-4)- | 158757-79-0 | 99.8% | 418.33 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD15705MG
Encompass_Preferred
BODIPYFL C12 is a BODIPY dye, a small molecule dye with strong ultraviolet absorption, a sharp fluorescence peak, and high quantum yield. It is relatively insensitive to environmental polarity and pH, and stable under various physiological conditions. Due to its structural asymmetry, it produces various structural products. BODIPY lipid droplet dyes can pass through the cell membrane, localize neutral lipids in the cell, and specifically stain lipid droplets for labeling live and fixed cells. Maximum excitation/emission wavelength: 480/508 nm.
  • Small molecule dye with strong ultraviolet absorption
  • Relatively sharp fluorescence peak
  • High quantum yield
  • Insensitive to environmental polarity and pH
  • Stable under various physiological conditions
  • Passes through cell membrane for cell labeling
  • Specifically stains lipid droplets
  • Suitable for labeling live and fixed cells
Catalog No. 50-004-30000