Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borate Buffer, pH 10.5, Ricca Chemical
SDP

$190.70 - $938.00

Specifications

Chemical Name or Material Borate Buffer
CAS 7732-18-5 , 1303-96-4 , 1310-73-2
Concentration 1X
For Use With (Application) Water Analysis
pH 10.5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
R1048000500
SDP
View Documents
Ricca Chemical Company
R1048000500A
500 mL Natural Poly Bottle
Each for $190.70
Add to Cart
 
R104800020F
SDP
View Documents
Ricca Chemical Company
R104800020F
20 L Cubitainer
Each for $938.00
Add to Cart
 
Description

Description

Specifications

Specifications

Borate Buffer
1X
10.5
Buffer
7732-18-5 , 1303-96-4 , 1310-73-2
Water Analysis
Liquid
Laboratory
Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.