Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borate Buffer, pH 10.5, Ricca Chemical
SDP

Catalog No. R104800020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R104800020F 20 L Cubitainer
R1048000500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R104800020F Supplier Ricca Chemical Company Supplier No. R104800020F
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Borate Buffer
CAS 7732-18-5, 1303-96-4, 1310-73-2
CAS Min % 97.0, 0.46, 0.09
Color Colorless
CAS Max % 100.0, 0.48, 0.09
Concentration 1X
For Use With (Application) Water Analysis
pH 10.5
Physical Form Liquid
Quantity 20 L
Solution Type Buffer
Packaging Cubitainer
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.