Learn More
ATCC Bordetella pertussis; genomic DNA
Supplier:  ATCC BAA589D5
Encompass
Genomic DNA isolated from Bordetella pertussis strain Tohama 1 (ATCC BAA-589). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-3819