Learn More
AG Scientific Inc Boric Acid ACS Grade 5 KG
Supplier:  AG Scientific Inc B26015KG
Encompass_Preferred
Boric Acid (also known as hydrogen borate) is a commonly used reagent in molecular biology for preparation of buffers. Boric acid shows some antibacterial and antifungal activity.
Catalog No. 50-226-8252