Learn More
Cayman Chemical -Bornyl Acetate 10g
SDP
Supplier:  Cayman Chemical 2961710 G
Encompass_Preferred
An acetate form of borneol; induces an EAG response in isolated P. americana antennae from both male and female cockroaches that is similar to the response induced by sex pheromones; induces hydrogen peroxide production and apoptosis in M. polymorpha plant cells
Catalog No. 50-299-2179