Learn More
Medchemexpress LLC Boronic acid, B-(3-aminophenyl)- | 30418-59-8 | MFCD00007755 | 99.9% | 136.94 g/mol | C6H8BNO2 | 100 G

Supplier: Medchemexpress LLC HYW017604100G
3-Aminophenylboronic acid (m-aminophenylboronic acid) is an organoboron compound used in research and synthesis. It serves as a versatile building block in organic chemistry and as a reagent for biochemical applications, including functionalization of nanomaterials for selective glycoprotein detection.
- Molecular formula C6H8BNO2 and molecular weight 136.94 g/mol.
- High purity suitable for analytical and synthetic use.
- Compatible with cross-coupling reactions such as Suzuki-Miyaura.
- Soluble in common organic solvents and available in solution forms.
- Supplied in multiple solid pack sizes for scale flexibility.
- Useful for derivatization and surface functionalization of nanomaterials.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.