Learn More
Bellco Glass, Inc. Borosil Allihn Condenser with I/C Inner & Outer Joint 300 MM
SDP
Supplier:  Bellco Glass, Inc. 452800717
Encompass_Preferred
Borosil Allihn Condenser with Drip Tip, Interchangeable Inner and Outer Joint, 300 mm, CS/5Borosil Allihn Condensers have an interchangeable inner and outer joint with a drip tip. The Allihn Condenser has a greater surface area than the Liebig, with a series of bulbs which generates a higher cooling capacity. This condenser is ideal for use in laboratory scale refluxing. There is an inlet and an outlet for attaching tubing to move coolant throughout the outer tube. Allihn condersers are used to keep liquids at their boiling temperature. Made of 3.3 borosilicate to withstand heat and temperature changes.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearApprox Jacket Length: 300mmApprox Overall Height: 450mmNeck Type: JointsInterchangeable Socket Joint: 24/ 29Interchangeable Cone Joint: 24/ 29Class/ Quality Grade: Type I, Class AQuantity Per Case: 5
Catalog No. 50-255-0045