Learn More
Bellco Glass, Inc. Borosil Beakers, Low-Form, with Spouts, 400mL
SDP
Supplier:  Bellco Glass, Inc. 452800912
Encompass_Preferred
Borosil Beaker Griffin Low Form with Spout Graduated ISO 3819 Borosilicate 400mL CS/40SpecificationsMaterial: GlassBeaker: 400mLApprox. O.D. x Height: 80mm x 110mmBorosilicate Glass 3.3Beakers are not calibrated. Calibration on beakers is for reference only. We can not provide a calibration certificate for beakers.
Catalog No. 50-254-9850