Learn More
Bellco Glass, Inc. Borosil Bottles, Reagent, Amber, with Stoppers, 250mL, 19/26
SDP
Supplier:  Bellco Glass, Inc. 452800703
Encompass_Preferred
Borosil Amber Reagent Bottles, Plain, Narrow Mouth, Graduated with Interchangeable Flat Head Solid Glass Stopper ISO 4796-2 Ground neck Borosilicate glass Durable Matt finish enamelled area for marking 250 mL CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 250 mLColor: AmberDimension: 70mm x 140mmGraduation Color: White EnamelClass/Quality Grade: Type 1, Class A
Catalog No. 50-255-0059