Learn More
Bellco Glass, Inc. Borosil Desiccators, Vacuum, 150mm (Small)
SDP
Supplier:  Bellco Glass, Inc. 452800742
Encompass_Preferred
Borosil Desiccator Vacuum, Stopcock with PTFE spindle and Porcelain plate, Borosilicate 3.3 glass, 100 mm, CS/1Borosil Vacuum Desiccators are generally used for the drying of moist products, and for the storage of moisture-sensitive substances to ensure sample stability. Each desiccator complies with IS 6128, coming equipped with bodies and covers that are manufactured to have great strength and utility. The plates are positioned on an internal ledge within the base. The base and cover are made of 3.3 borosilicate glass and for your convenience, all Borosil brand Plain Desiccators of the same size have interchangeable bodies and covers. This type is specifically for use under a vacuum and comes with a glass stopcock at the top.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearApprox ID Ground Flange: 100mmClass/ Quality Grade: Type IQuantity Per Case: 1Steam Sterilization at 121º C is recommended for autoclavingNot recommended for heat applications
Catalog No. 50-255-0020