Learn More
Bellco Glass, Inc. Borosil Flasks, Volumetric, Class A, WM, Amber, Glass St., 50mL, 14/23, Bat. Cert
SDP
Supplier:  Bellco Glass, Inc. 452801059
Encompass_Preferred
DescriptionBorosil Wide Mouth Volumetric Flask with Interchangeable Solid Glass Stopper, Accuracy as per Class A, ASTM Standard with Batch Certificate, 50mL, Borosilicate 3.3 CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Bottle volume: 50mLStopper Size: 14/23Color: AmberTolerance: +/- 0.08Overall Dimension:11.81" L x 8.27" W x 5.12" HClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9703