Learn More
Bellco Glass, Inc. Borosil Flasks, Volumetric, Class A, WM, Amber, Glass St., 100mL, 19/26, Ind. Cert
SDP
Supplier:  Bellco Glass, Inc. 452801062
Encompass_Preferred
Borosil Flasks, Volumetric, Class A, WM, Amber, Glass St., 100mL, 19/26, Ind. Cert
Catalog No. 50-254-9700