Learn More
Bellco Glass, Inc. Borosil Funnels Short Stem Plain 60deg 25 MM ISO 4798 Borosilicate
Supplier: Bellco Glass, Inc. 452800829
Borosil Funnels, Short Stem, Plain, 60-degree angle, DIN/ISO 4798, 25mm Diameter, CS/30Borosil Funnels may be used to separate solids from liquids, liquids from liquids, and occasionally for pouring something into a container. The short stem funnels comply with IS 1591, DIN/ ISO 4798, and are manufactured for high thermal shock resistance and chemical resistance. They are also designed with a 60 bowl angle to ensure the precise fitting of the filter paper. Generally, short-stemmed funnels are used for filtering and decanting liquids of varied densities.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearDiameter: 25mmClass/ Quality Grade: Type IQuantity Per Case: 30