Learn More
Bellco Glass, Inc. Borosil Pipettes, Measuring (Mohr), Class A, 5.0mL x 0.05mL, Individual Cert
SDP
Supplier:  Bellco Glass, Inc. 452801076
Encompass_Preferred
DescriptionBorosil Graduated Mohr Pipette Class A Individual Certificate, ISO 835, 3.3 Borosilicate glass USP Type I, Amber markings, 5 mL, CS/10Borosil Pipettes, measuring (Mohr Type), Class A with Individual Certificate comply with IS, EN ISO 835. These Pipettes are graduated for delivery from the zero mark to the last graduation mark. Built sturdy with a design for long lasting quality. The markings are made by permanent amber stain diffused into the surface of the glass for visibility and durability. Borosil brand Pipettes are truly dependable in terms of their accuracy.SpecificationsMaterial: 3.3 Borosilicate GlassColor: ClearCapacity: 5 mLGraduation Rangel: 0 - 5 mLGraduation Interval: 0.05 mLGraduation Color: AmberClass/ Quality Grade: Type I, Class ATolerance: 0.03 mLQuantity Per Case: 10
Catalog No. 50-254-9686