Learn More
Bellco Glass, Inc. Borosil Pipettes, Serological, Class A, 1mL x 0.01mL, Batch Cert
SDP
Supplier:  Bellco Glass, Inc. 452801111
Encompass_Preferred
Borosil Graduated Serological pipettes Class A, ASTM 1293-02, Batch Certificate, 3.3 Borosilicate, To Deliver, CS/10, 1mL(1*0.01)SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 1mLOverall Dimension: 14.96" L x 14.96" W x 5.12" HGrad. Interval: .01Tolerance: .01Class/Quality Grade: Type 1, Class A
Catalog No. 50-254-9651