Learn More
Bellco Glass, Inc. Borosil Pipettes, Volumetric, Class A, 3mL, Batch Cert
SDP
Supplier:  Bellco Glass, Inc. 452801127
Encompass_Preferred
Borosil Volumetric Transfer Pipette Class A Accuracy ASTM E-969-02 with Batch Certificate 2mL Borosilicate 3.3, CS/10SpecificationsMaterial: 3.3 Borosilicate Glass Color: ClearCapacity: 3mLOverall Dimension: 14.96" L x 5.12" W x 2.17" HTolerance: .01mLClass/Quality Grade: Type 1, Class A
Catalog No. 50-254-9635