Learn More
Bellco Glass, Inc. Borosil Plastic PP Stopper For Vol. Flasks 7/16
SDP
Supplier:  Bellco Glass, Inc. 452800866
Encompass_Preferred
Borosil Plastic Polypropylene Stopper for Volumetric Flasks, Interchangeable 7/16, CS/100SpecificationsMaterial: PolypropyleneColor: ClearInterchangeable: 7/16Class/ Quality Grade: Class VINeck Type: StopperQuantity Per Case: 100
Catalog No. 50-254-9896