Learn More
Bellco Glass, Inc. Borosil Tubes, Test, Reusable, Plain End, 20mL, 16mm x 150mm (OD x H)
SDP
Supplier:  Bellco Glass, Inc. 452801215
Encompass_Preferred
Test Tubes without RIM 16*150 Borosilicate 3.3 ISO 4142 20ml CS/400SpecificationsMaterial: GlassVolumen: 20 mLOverall Dimension: 16mm x 150mmBorosilicate Glass 3.3Graduation Range: Type I
Catalog No. 50-254-9547