Learn More
Bellco Glass, Inc. Borosil Culture Tubes Media Flat Bottom PP cap with Liner 5 mL
SDP
Supplier:  Bellco Glass, Inc. 452800895
Encompass_Preferred
Borosil Media Culture Tubes, Flat Bottom, PP Screw Cap with PTFE Liner, 5 mL, CS/100Borosil Media Culture Tubes with a Flat Bottom are the ideal replacement for universal bottles. The flat bottom design is convenient for storage. The polypropylene screw cap has a PTFE liner to help seal and secure the inner contents inside the culture tubes. The caps are specially designed to facilitate the handling and sealing when in use. Material: 3.3 Borosilicate GlassColor: ClearCapacity: 5mLApprox O.D. x Length: 16mm x 50mmClass/ Quality Grade: Type I, Class AQuantity Per Case: 100
Catalog No. 50-254-9867