Learn More
Chemglass Life Sciences Chemglass Borosilicate Tubing for Swagelok compression fittings - 3/4 in OD ±0.002 in, 15.0 mm ID, 14-3/4 in length (CG-713-04)
SDP
Supplier:  Chemglass Life Sciences CG71304
Encompass_Preferred
Chemglass CG-713-04 is a laboratory-grade borosilicate glass tube engineered for compression fittings, providing precise dimensions and reliable sealing for fluid-transfer applications.
  • Made from borosilicate glass for chemical resistance and thermal stability.
  • Outer diameter 3/4 in ±0.002 in for consistent compression sealing.
  • Inner diameter 15.0 mm to accommodate specified flow requirements.
  • Length 14-3/4 in for standard installation spans.
  • Compatible with PTFE ferrules to improve sealing and chemical compatibility.
Catalog No. 50-273-3516