Learn More
Medchemexpress LLC HY-10227 200mg , Bortezomib PS-341;Brotezamide;DPBA;LDP 341;MG 341;Radiciol;NSC 681239 CAS:179324-69-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10227200MG
Encompass_Preferred
Medchemexpress, HY-10227 200mg Bortezomib PS-341;Brotezamide;DPBA;LDP 341;MG 341;Radiciol;NSC 681239 CAS:179324-69-7 Formula:C19H25BN4O4 Ki: 0.6 nM (20S proteasome) Purity:98% Bortezomib is a potent 20S proteasome inhibitor with K i of 0.6 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2520