Learn More
Medchemexpress LLC Bortezomib-d8 | 98.0% | 392.29 g/mol | C19H17D8BN4O4 | 100 UG
SDP
Supplier:  Medchemexpress LLC HY10227S100UG
Encompass_Preferred
Bortezomib-d8 is a deuterium-labeled analogue of the proteasome inhibitor bortezomib supplied as an isotope-labeled reference standard for analytical and research use. It is intended for mass spectrometry, pharmacokinetic, and mechanism-of-action studies where a labeled internal standard is required. The compound has molecular formula C19H17D8BN4O4, molecular weight 392.29 g/mol, and is supplied with reported chemical purity of 98.0%.
  • Deuterium-labeled internal standard for mass spectrometry applications.
  • High chemical purity (98.0%) for reliable analytical results.
  • Supplied in small research quantities suitable for analytical workflows.
  • Soluble in DMSO and ethanol with reported concentrations useful for stock preparation.
  • Stable when stored at -20°C (solid) and for defined periods at -80°C in solution.
Catalog No. 50-002-46352