Learn More
Cayman Chemical Bortezomib Inhibitors
SDP
Supplier:  Cayman Chemical 100088225 MG
Encompass_Preferred
A dipeptide boronic acid derivative that reversibly inhibits the 20S proteasome (Ki = 0.6 nM); drives cell cycle arrest and apoptosis in cancer cell lines and has applications in multiple myeloma and certain lymphomas, as well as other types of cancer
Catalog No. 50-288-9134