Learn More
Selleck Chemical LLC Bosentan Hydrate-10mg
SDP
Supplier:  Selleck Chemical LLC S305110MG
Encompass_Preferred
Bosentan (Ro 47-0203) is an endothelin (ET) receptor antagonist for ET-A and ET-B with Ki of 4.7 nM and 95 nM, respectively.
Catalog No. 50-238-9638