Learn More
Medchemexpress LLC Bosentan hydrate | 157212-55-0 | 99.9% | C27H31N5O7S | 100 MG
SDP
Supplier:  Medchemexpress LLC HYA0013A100MG
Encompass_Preferred
Bosentan hydrate is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors, with Ki values of 4.7 nM and 95 nM in human SMC, respectively. It has a molecular weight of 569.63 g/mol.
  • Competitive and dual antagonist of endothelin-1 (ET)
  • Targets ETA and ETB receptors
  • Ki of 4.7 nM for ETA and 95 nM for ETB in human SMC
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-004-32984