Learn More
Medchemexpress LLC HY-A0013A 500mg Medchemexpress, Bosentan (hydrate) CAS:157212-55-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0013A500MG
Encompass_Preferred
Medchemexpress, HY-A0013A 500mg Bosentan (hydrate) CAS:157212-55-0 Bosentan hydrate is a competitive and dual antagonist of endothelin-1 (ET) for the ETA and ETB receptors with Ki of 4.7 nM and 95 nM in human SMC, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9376