Learn More
AdipoGen Bostrycin
SDP
Supplier:  AdipoGen AGCN20175C250
Encompass_Preferred
Chemical. CAS 21879-81-2. Formula C16H16O8. MW 336.3. Isolated from fungus Arthrinium sp. Anthraquinone antibiotic. Red antibacterial agent against Gram positive bacteria. Anticancer agent. Inhibits cell proliferation of human lung carcinoma A549 cells via downregulation of the PI3K/Akt pathway. Also shown to block cell cycle at G1 phase leading to cell death. Induced mitochondria-mediated apoptosis in the yeast Saccharomyces cerevisiae. Weak in vitro inhibitor of Mycobacterium tuberculosis protein tyrosine phosphatase MptpB. Potential red-colored agent for food processing.
Catalog No. 50-256-2641