Learn More
Medchemexpress LLC β-Boswellic acid | 631-69-6 | MFCD04039448 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN25131MG
Encompass_Preferred
β-Boswellic acid is isolated from the gum resin of *Boswellia serrata*. It exhibits anticancer, antioxidant, anti-inflammatory, and anti-arthritic pain activities. This compound is an orally active, nonreducing-type inhibitor of 5-lipoxygenase (5-LO) product formation, working either by directly interacting with 5-LO or by blocking its translocation. It inhibits the synthesis of DNA, RNA, and protein in human leukemia HL-60 cells. It shows promise for research in diabetes, inflammatory, and arthritic diseases.
  • Exhibits anticancer activity
  • Offers antioxidant benefits
  • Provides anti-inflammatory effects
  • Alleviates anti-arthritic pain
  • Orally active 5-lipoxygenase (5-LO) inhibitor
  • Inhibits DNA, RNA, and protein synthesis in HL-60 cells
  • Promising for diabetes, inflammatory, and arthritic disease research
Catalog No. 50-004-21287