Learn More
Hach Company Brush, Bottle
Supplier:  Hach Company 2929601
Encompass
The pivot head on this brush swivels, enabling you to thoroughly clean bottles and flasks, including difficult curves and corners. Natural bristles set in wood will not scratch glass. Uniquely designed head fits through a 29/26 standard taper neck or any neck with a diameter of 24mm (1") or more. The brush has a 45mm (11⁄4") wooden back and a 33cm (13") long hardwood handle.
Catalog No. 50-199-6829