Learn More
Wilmad Labglass BOTTLE GAS WASH 125ML COMP C
Supplier:  Wilmad Labglass LG3765124
Encompass_Preferred
Bottle, Gas Washing, Tall Form, with Fritted Disc. Capacity: 125mL, Joint Size: [ST] 40/35; Porosity: Coarse. Manufactured with Inlet tube sealed through center of fritted glass disc. Inlet tube extends nearly to base of bottle.
Catalog No. 50-804-539
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.