Learn More
Wilmad Labglass BOTTLE GAS WASH 250ML COMP C
Supplier:  Wilmad Labglass LG3765126
Encompass_Preferred
Bottle, Gas Washing, Tall Form, with Fritted Disc. Capacity: 250mL, Joint Size: [ST] 40/35; Porosity: Coarse. Manufactured with Inlet tube sealed through center of fritted glass disc. Inlet tube extends nearly to base of bottle.
Catalog No. 50-803-890