Learn More
Wilmad Labglass BOTTLE GAS WASH FRTD CYL 250ML
Supplier:  Wilmad Labglass LG3761102
Encompass_Preferred
Bottle, Gas Washing, Tall Form, with Fritted Cylinder. Complete. Capacity: 250mL, Joint Size: [ST] 29/42; Porosity: Coarse. Manufactured with 12mm OD x 25mm length fritted cylinder. Supplied with LG-1004-110 plastic joint clamp.
Catalog No. 50-806-527
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.