Learn More
Chemglass Life Sciences Chemglass Media Bottle, Square, Sterile, 125 mL (Nest Pack)
SDP
Supplier:  Chemglass Life Sciences CGN1151125
Encompass_Preferred
Sterile 125 mL square media bottle for storage and handling of culture media, buffers, and reagents. The nest-packed bottles include graduations for approximate volume measurement and a leak-resistant HDPE cap for secure sealing.
  • 125 mL capacity
  • Sterile, gamma-irradiated
  • Square profile for space-efficient storage
  • Graduated scale for approximate volume reading
  • Packaged in nest packs (6 per pack, 48 per case)
  • High-density polyethylene (HDPE) cap for leak resistance
  • DNase-, RNase-, and pyrogen-free
  • Good chemical resistance and break-resistant construction
Catalog No. 50-273-3506