Learn More
Teledyne ISCO, Inc BOTTLE RACK FOR TORRENT
Supplier:  Teledyne ISCO, Inc 605247004
Encompass
Set of two racks for (18) 480 mL glass bottles. Each rack holds (9) bottles. Fits Foxy R2 only; requires qty. 1 set. 500 mL French Square, 68 mm on a side
Catalog No. NC9032984