Learn More
Hach Company Bottle, Wash, Nalgene, Narrow Mouth, 500 mL, Blue Cap/Stem, 6/pk
Supplier:  Hach Company 2928202
Encompass
Narrow mouth polypropylene screw closure and stem feature one-piece construction to ensure a leak proof seal. Tip can be cut back to increase flow. 500mL (16.8 oz. Capacity, 28mm mouth). Choose from 4 cap/stem colors.
Catalog No. 50-199-6807