Learn More
Electron Microscopy Sciences Bouin's Fixative 10 ML
SDP
Supplier:  Electron Microscopy Sciences 15990
Encompass_Preferred
Bouin's Fixative is used in light microscopy for the preparation of delicate structure. Prepared fresh upon receipt of order to ensure maximum shelf life. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
Catalog No. 50-247-86