Learn More
ABclonal Technology BOULE/BOLL Rabbit mAb
SDP
Supplier:  ABclonal Technology A2766320UL
Encompass_Preferred
This gene belongs to the DAZ gene family required for germ cell development It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like) Loss of this gene function results in the absence of sperm in semen (azoospermia) Histological studies demonstrated that the primary defect is at the meiotic G2 M transition Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene
Catalog No. 50-000-08894