Learn More
Cayman Chemical BovIn BSA10% 15mL
SDP
Supplier:  Cayman Chemical 70191715 ML
Encompass_Preferred
BSA (10%) has been tested and formulated to work exclusively with SARS-CoV-2 Spike S1 RBD-ACE2 Binding Cellular Imaging Assay Kit
Catalog No. 50-289-3813