Learn More
Immunology Consultants Bovine Alpha 1-Acid Glycoprotein (AGP) ELISA Kit
SDP
Supplier:  Immunology Consultants E10AGP
Encompass_Preferred
The Bovine AGP test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring AGP in bovine biological samples.
Catalog No. 50-240-6579