Learn More
American Research Products Inc BoVine anti Mullerian hormone
Supplier:  American Research Products Inc CSBE13406B
Encompass
BoVine anti-Mullerian hormone (AMH) ELISA kit from CUSABIO Detection Range 0 5 ng/mL-200 ng/mL Sensitivity 0 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29426