Learn More
American Research Products Inc BoVine C reactiVe protein CRP
Supplier:  American Research Products Inc CSBE08577B
Encompass
BoVine C-reactiVe protein CRP ELISA Kit from CUSABIO Detection Range 1 25 ng/mL-80 ng/mL Sensitivity 1 25 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29114