Learn More
Biomatik Corporation Bovine Fibroblast Growth Factor 4 (FGF4) ELISA Kit
Supplier:  Biomatik Corporation EKU04167
Encompass_Preferred
Bovine Fibroblast Growth Factor 4 (FGF4) ELISA Kit for detecting FGF4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.62-1000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48803. Target Synonyms: HBGF4; HST; HSTF1; KFGF; K-FGF; Heparin Secretory Transforming Protein 1; Kaposi Sarcoma Oncogene; Transforming Protein KS3; FGF-Related Oncogene. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6838