Learn More
Bioworld Bovine gamma globulin, 10 mL
Supplier:  Bioworld 220700021
Encompass_Preferred
Bovine γ- Globulin Standard (2mg/ml)SpecificationPrediluted protein standard2mg/ml in saline bufferSupplied as 5x2mL sizeshipped ambient
Catalog No. 50-253-887