Learn More
Immunology Consultants Bovine IgG ELISA Kit - (Min. Reactivity With Hu,Ms,Rb,Rt)
SDP
Supplier:  Immunology Consultants E10GXSPP
Encompass_Preferred
This Bovine IgG ELISA kit is intended for the quantitative determination of total bovine IgG. There is no reactivity to human, mouse, rabbit and rat immunoglobulins. The IgG test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA).
Catalog No. 50-240-6524