Learn More
Immunology Consultants Bovine IgG ELISA Kit
SDP
Supplier:  Immunology Consultants E10G
Encompass_Preferred
The Bovine IgG ELISA kit is intended for the quantitative determination of total bovine IgG in biological samples. The IgG test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA).
Catalog No. 50-240-6583