Learn More
American Research Products Inc BoVine Interleukin 1b IL 1b
Supplier:  American Research Products Inc CSBE12986B
Encompass
BoVine Interleukin 1b (IL- 1b) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29945