Learn More
Haematologic Technologies Inc Bovine Lactadherin, 50 ug
Supplier:  Haematologic Technologies Inc BLAC1200
Encompass_Preferred
Purified lactadherin functions as an anticoagulant by blocking phosphatidylserine-containing membrane sites for blood coagulation proteins. Fluoresence-labeled lactadherin functions as a sensitive probe for exposed phosphatidylserine on nucleated cells and on stimulated platelets. Lactadherin will bind to membranes that have phosphatidylserine content below the threshold for annexin V binding.
Catalog No. NC0257008