Learn More
Sigma Aldrich Fine Chemicals Biosciences Bovine Serum Albumin, 9048-46-8, MFCD00130384, 10g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A450310G
Encompass
Cold ethanol fraction, pH 5.2, ≥96%, Synonym: Albumin bovine serum, BSA, Bovine albumin; Bovine serum albumin is a model protein in various studies and is made up of 583 amino acids.
Catalog No. NC1639690